Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692673.3 | XP_005692730.1 | 2 - 3 | 203 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |