Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697844.3 | XP_005697901.1 | 2 - 3 | 1062 | 89 | |
XM_005697845.3 | XP_005697902.1 | 2 - 3 | 895 | 89 | ||
XM_005697846.3 | XP_005697903.1 | 2 - 3 | 580 | 89 | ||
XM_005697847.3 | XP_005697904.1 | 2 - 3 | 572 | 89 | ||
XM_005697848.3 | XP_005697905.1 | 2 - 3 | 1059 | 89 | ||
XM_005697850.3 | XP_005697907.1 | 2 - 3 | 1063 | 89 | ||
XM_013974681.2 | XP_013830135.1 | 2 - 3 | 565 | 89 | ||
XM_013974683.2 | XP_013830137.1 | 2 - 3 | 892 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |