Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689597.1 | XP_005689654.1 | 2 - 3 | 210 | 75 | |
XM_018058986.1 | XP_017914475.1 | 2 - 3 | 185 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |