Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692748.3 | XP_005692805.2 | 4 - 5 | 677 | 112 | |
XM_018062869.1 | XP_017918358.1 | 3 - 4 | 479 | 112 | ||
GenBank mRNA | JO367733.2 | - | 3 - 4 | 873 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |