Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058765.1 | XP_017914254.1 | 1 | 1333 | 111 | |
XM_018058766.1 | XP_017914255.1 | 1 | 1333 | 111 | ||
XM_018058767.1 | XP_017914256.1 | 2 | 302 | 111 | ||
XM_018058768.1 | XP_017914257.1 | 2 | 169 | 111 | ||
XM_018058769.1 | XP_017914258.1 | 2 | 193 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |