Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062862.1 | XP_017918351.1 | 1 - 2 | 564 | 54 | |
GenBank mRNA | JO440462.2 | - | 3 - 4 | 1025 | 54 | |
KA322134.1 | - | 1 - 2 | 350 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |