Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172986.1 | NP_001166457.1 | 2 - 3 | 236 | 71 | |
XM_005004596.3 | XP_005004653.1 | 2 - 3 | 261 | 71 | ||
GenBank mRNA | AB055403.1 | - | 2 - 3 | 236 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Cavia porcellus Probe spans exons |