Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173003.1 | NP_001166474.1 | 4 - 5 | 548 | 66 | |
XM_005003770.3 | XP_005003827.2 | 4 - 5 | 949 | 66 | ||
XM_013152814.2 | XP_013008268.1 | 3 - 4 | 800 | 66 | ||
GenBank mRNA | AF088283.1 | - | 4 - 5 | 545 | 66 | |
AF088284.1 | - | 4 - 5 | 548 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Cavia porcellus Probe spans exons |