Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173013.1 | NP_001166484.1 | 3 - 4 | 1042 | 78 | |
XM_013144836.2 | XP_013000290.1 | 4 - 5 | 1125 | 78 | ||
GenBank mRNA | M57269.1 | - | 3 - 4 | 1042 | 78 |
Gene Symbol: Apf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005000487.3 | XP_005000544.1 | 4 - 5 | 1100 | 78 | |
GenBank mRNA | M57271.1 | - | 3 - 4 | 1022 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |