Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173033.1 | NP_001166504.1 | 547 | 66 | ||
GenBank mRNA | EU664998.1 | - | 547 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Cavia porcellus |