Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173040.1 | NP_001166511.1 | 2 - 3 | 740 | 94 | |
GenBank mRNA | EU827601.1 | - | 2 - 3 | 740 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |