Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grifin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173083.1 | NP_001166554.1 | 2 - 3 | 96 | 87 | |
GenBank mRNA | EB370872.1 | - | 2 - 3 | 96 | 87 | |
EU259197.1 | - | 2 - 3 | 96 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Cavia porcellus Probe spans exons |