Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173401.1 | NP_001166872.1 | 2 - 3 | 387 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |