Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169651.2 | NP_001163122.1 | 4 - 5 | 929 | 75 | |
NM_001169652.2 | NP_001163123.1 | 5 - 6 | 1140 | 75 | ||
NM_136867.3 | NP_610711.1 | 5 - 6 | 1140 | 75 | ||
NM_165852.2 | NP_725088.1 | 4 - 5 | 929 | 75 | ||
GenBank mRNA | AY122187.1 | - | 5 - 6 | 1104 | 75 | |
BT088945.1 | - | 4 - 5 | 936 | 75 | ||
EF587234.1 | - | 4 - 5 | 929 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |