Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Den1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169655.2 | NP_001163126.1 | 2 | 1082 | 133 | |
NM_136919.3 | NP_610763.1 | 2 | 1178 | 133 | ||
GenBank mRNA | AY119635.1 | - | 2 | 1165 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |