Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir76a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104177.3 | NP_001097647.3 | 6 - 7 | 958 | 96 | |
NM_001144510.3 | NP_001137982.1 | 4 - 5 | 702 | 96 | ||
NM_140891.4 | NP_649148.2 | 4 - 5 | 689 | 96 | ||
NM_168817.3 | NP_730452.2 | 5 - 6 | 728 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |