Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitslre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140994.4 | NP_649251.2 | 1 - 2 | 336 | 57 | |
NM_168869.3 | NP_730563.1 | 2 - 3 | 277 | 57 | ||
GenBank mRNA | BT016092.1 | - | 1 - 2 | 327 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |