Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACXE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144344.3 | NP_652601.3 | 6 - 7 | 1611 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |