Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058010.4 | NP_477358.1 | 6 - 7 | 1498 | 90 | |
NM_166102.2 | NP_725476.1 | 7 - 8 | 1510 | 90 | ||
GenBank mRNA | AY058794.1 | - | 6 - 7 | 1473 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |