Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080084.3 | NP_524823.1 | 6 - 7 | 2018 | 76 | |
NM_166144.2 | NP_725530.1 | 6 - 7 | 2018 | 76 | ||
NM_166145.2 | NP_725531.1 | 6 - 7 | 2018 | 76 | ||
NM_166146.2 | NP_725532.1 | 6 - 7 | 2018 | 76 | ||
NM_166147.2 | NP_725533.1 | 5 - 6 | 1756 | 76 | ||
GenBank mRNA | AF212366.1 | - | 6 - 7 | 1981 | 76 | |
AF212367.1 | - | 6 - 7 | 1981 | 76 | ||
AF212368.1 | - | 6 - 7 | 1981 | 76 | ||
AF212369.1 | - | 6 - 7 | 1981 | 76 | ||
BT032842.1 | - | 6 - 7 | 1980 | 76 | ||
BT150099.1 | - | 6 - 7 | 1974 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |