Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AcCoAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014599.2 | NP_001014599.2 | 1 - 2 | 504 | 110 | |
NM_168894.3 | NP_730611.1 | 1 - 2 | 766 | 110 | ||
GenBank mRNA | BT126330.1 | - | 1 - 2 | 766 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |