Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043154.4 | NP_001036619.1 | 6 - 7 | 1372 | 84 | |
NM_001170022.3 | NP_001163493.1 | 6 - 7 | 1360 | 84 | ||
NM_001259944.2 | NP_001246873.1 | 7 - 8 | 1472 | 84 | ||
NM_001259945.2 | NP_001246874.1 | 6 - 7 | 1360 | 84 | ||
NM_001259946.2 | NP_001246875.1 | 6 - 7 | 1372 | 84 | ||
NM_001259947.1 | NP_001246876.1 | 7 - 8 | 1484 | 84 | ||
NM_001275268.1 | NP_001262197.1 | 7 - 8 | 1472 | 84 | ||
NM_001275269.1 | NP_001262198.1 | 7 - 8 | 1431 | 84 | ||
NM_001316446.1 | NP_001303375.1 | 6 - 7 | 1372 | 84 | ||
NM_079484.6 | NP_524208.1 | 6 - 7 | 1372 | 84 | ||
NM_168946.4 | NP_730700.2 | 7 - 8 | 1472 | 84 | ||
NM_206423.5 | NP_996145.1 | 6 - 7 | 1372 | 84 | ||
GenBank mRNA | AY060403.1 | - | 6 - 7 | 1320 | 84 | |
BT030417.1 | - | 6 - 7 | 1296 | 84 | ||
X99340.1 | - | 2127 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |