Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc68Ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206332.2 | NP_996054.1 | 9389 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |