Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042794.4 | NP_001036259.2 | 5 - 6 | 1485 | 72 | |
NM_001169178.1 | NP_001162649.1 | 6 - 7 | 1110 | 72 | ||
NM_001169179.1 | NP_001162650.1 | 5 - 6 | 732 | 72 | ||
NM_001272263.1 | NP_001259192.1 | 6 - 7 | 2556 | 72 | ||
NM_001272264.1 | NP_001259193.1 | 6 - 7 | 2687 | 72 | ||
NM_057366.5 | NP_476714.1 | 5 - 6 | 1117 | 72 | ||
NM_057367.5 | NP_476715.1 | 5 - 6 | 756 | 72 | ||
NM_057368.5 | NP_476716.2 | 4 - 5 | 2714 | 72 | ||
NM_134278.3 | NP_599105.1 | 5 - 6 | 1019 | 72 | ||
NM_166947.3 | NP_726822.1 | 5 - 6 | 964 | 72 | ||
NM_166948.4 | NP_726823.1 | 5 - 6 | 1117 | 72 | ||
NM_206611.2 | NP_996334.1 | 4 - 5 | 673 | 72 | ||
NM_206612.3 | NP_996335.1 | 4 - 5 | 1102 | 72 | ||
NM_206613.3 | NP_996336.1 | 7 - 8 | 1406 | 72 | ||
NM_206614.3 | NP_996337.1 | 6 - 7 | 1000 | 72 | ||
NM_206615.2 | NP_996338.1 | 5 - 6 | 930 | 72 | ||
GenBank mRNA | AY122193.1 | - | 5 - 6 | 1092 | 72 | |
BT050474.1 | - | 4 - 5 | 1052 | 72 | ||
BT072831.1 | - | 5 - 6 | 741 | 72 | ||
BT133153.1 | - | 4 - 5 | 2708 | 72 | ||
X70863.1 | - | 1076 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |