Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103415.2 | NP_001096885.2 | 9 - 10 | 1707 | 82 | |
NM_001272322.1 | NP_001259251.1 | 12 - 13 | 3135 | 82 | ||
NM_001272323.2 | NP_001259252.1 | 11 - 12 | 3033 | 82 | ||
NM_001272324.2 | NP_001259253.1 | 11 - 12 | 3033 | 82 | ||
NM_001297957.1 | NP_001284886.1 | 9 - 10 | 1952 | 82 | ||
NM_001297958.1 | NP_001284887.1 | 11 - 12 | 3033 | 82 | ||
NM_001297959.1 | NP_001284888.1 | 9 - 10 | 1946 | 82 | ||
NM_001297960.1 | NP_001284889.1 | 11 - 12 | 3036 | 82 | ||
NM_080339.3 | NP_525078.3 | 9 - 10 | 1946 | 82 | ||
NM_167029.3 | NP_726986.3 | 11 - 12 | 3036 | 82 | ||
NM_167030.2 | NP_726987.2 | 9 - 10 | 1946 | 82 | ||
NM_167032.2 | NP_726989.3 | 9 - 10 | 1952 | 82 | ||
NM_206631.4 | NP_996354.2 | 11 - 12 | 3036 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |