Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144728.3 | NP_001138200.1 | 9 - 10 | 1791 | 74 | |
NM_001258775.3 | NP_001245704.1 | 9 - 10 | 1791 | 74 | ||
NM_001258776.2 | NP_001245705.1 | 10 - 11 | 1782 | 74 | ||
NM_001272661.2 | NP_001259590.1 | 10 - 11 | 1990 | 74 | ||
NM_001272662.2 | NP_001259591.1 | 10 - 11 | 1990 | 74 | ||
NM_001298382.1 | NP_001285311.1 | 9 - 10 | 2473 | 74 | ||
NM_078634.7 | NP_523358.5 | 9 - 10 | 2473 | 74 | ||
NM_206752.3 | NP_996475.2 | 9 - 10 | 1791 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |