Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 13 - 14 | 5634 | 65 | |
NM_140031.3 | NP_648288.2 | 12 - 13 | 6873 | 65 | ||
NM_168314.3 | NP_729451.1 | 3 - 4 | 2847 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |