Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FoxK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274772.2 | NP_001261701.1 | 4 - 5 | 1532 | 92 | |
NM_001274774.2 | NP_001261703.1 | 4 - 5 | 1532 | 92 | ||
NM_140183.3 | NP_648440.2 | 4 - 5 | 1532 | 92 | ||
NM_168443.3 | NP_729672.1 | 4 - 5 | 1514 | 92 | ||
NM_168444.2 | NP_729673.2 | 4 - 5 | 1613 | 92 | ||
NM_168445.3 | NP_729674.2 | 4 - 5 | 1410 | 92 | ||
NM_168446.2 | NP_729675.2 | 4 - 5 | 1488 | 92 | ||
GenBank mRNA | AY787838.1 | - | 3 - 4 | 1340 | 92 | |
BT023507.1 | - | 4 - 5 | 1468 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |