Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316387.1 | NP_001303316.1 | 3 - 4 | 2235 | 105 | |
NM_164818.2 | NP_723391.2 | 3 - 4 | 2235 | 105 | ||
GenBank mRNA | AF179360.1 | - | 2 - 3 | 2101 | 105 | |
AJ249178.1 | - | 2 - 3 | 1864 | 105 | ||
AY089558.1 | - | 3 - 4 | 2219 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |