Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRalpha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273373.2 | NP_001260302.1 | 4 - 5 | 2373 | 61 | |
NM_164874.3 | NP_723494.2 | 4 - 5 | 2579 | 61 | ||
NM_205952.3 | NP_995674.1 | 4 - 5 | 2579 | 61 | ||
NM_205953.3 | NP_995675.1 | 3 - 4 | 1449 | 61 | ||
GenBank mRNA | AF321445.1 | - | 2 - 3 | 586 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |