Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135072.2 | NP_608916.1 | 4 - 5 | 622 | 74 | |
GenBank mRNA | AY051602.1 | - | 4 - 5 | 622 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |