Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140385.4 | NP_648642.1 | 5 - 6 | 1671 | 77 | |
NM_168543.2 | NP_729891.1 | 5 - 6 | 1606 | 77 | ||
GenBank mRNA | AY061340.1 | - | 5 - 6 | 1652 | 77 | |
BT099796.1 | - | 5 - 6 | 1606 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |