Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300136.1 | NP_001287065.1 | 4 - 5 | 2700 | 71 | |
NM_140415.4 | NP_648672.1 | 4 - 5 | 2700 | 71 | ||
GenBank mRNA | AY051947.1 | - | 4 - 5 | 2664 | 71 |
Gene Symbol: Nxf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300135.1 | NP_001287064.2 | 3 - 4 | 5716 | 71 | |
NM_168566.2 | NP_729938.2 | 3 - 4 | 5716 | 71 | ||
GenBank mRNA | BT021396.1 | - | 3 - 4 | 5690 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |