Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yuri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316397.1 | NP_001303326.1 | 8 - 9 | 1785 | 61 | |
NM_165107.3 | NP_723904.1 | 7 - 8 | 2128 | 61 | ||
NM_165109.3 | NP_723906.1 | 8 - 9 | 2073 | 61 | ||
GenBank mRNA | BT001564.1 | - | 7 - 8 | 2126 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |