Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca-alpha1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273551.1 | NP_001260480.2 | 30 - 31 | 7933 | 78 | |
NM_001273552.1 | NP_001260481.1 | 29 - 30 | 6846 | 78 | ||
NM_001273554.1 | NP_001260483.1 | 29 - 30 | 6846 | 78 | ||
NM_001273555.1 | NP_001260484.1 | 29 - 30 | 6855 | 78 | ||
NM_001273556.1 | NP_001260485.1 | 30 - 31 | 6954 | 78 | ||
NM_080365.3 | NP_525104.3 | 30 - 31 | 7938 | 78 | ||
NM_134429.3 | NP_602305.2 | 30 - 31 | 7929 | 78 | ||
NM_165146.2 | NP_723952.2 | 31 - 32 | 7951 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |