Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP-2alpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057471.6 | NP_476819.2 | 8 - 9 | 3277 | 55 | |
NM_205885.2 | NP_995607.1 | 7 - 8 | 2844 | 55 | ||
GenBank mRNA | BT032839.1 | - | 8 - 9 | 3258 | 55 | |
BT125866.1 | - | 7 - 8 | 2844 | 55 | ||
Y11104.1 | - | 3146 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |