Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nompC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103619.4 | NP_001097089.2 | 6 - 7 | 1664 | 58 | |
NM_001258962.3 | NP_001245891.1 | 6 - 7 | 1664 | 58 | ||
NM_001316380.1 | NP_001303309.1 | 6 - 7 | 1664 | 58 | ||
NM_078759.4 | NP_523483.2 | 6 - 7 | 1664 | 58 | ||
NM_205912.4 | NP_995634.2 | 6 - 7 | 1664 | 58 | ||
GenBank mRNA | HM582118.1 | - | 6 - 7 | 1006 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |