Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273397.1 | NP_001260326.1 | 6 - 7 | 2598 | 103 | |
NM_001273398.1 | NP_001260327.1 | 6 - 7 | 2418 | 103 | ||
NM_135539.3 | NP_609383.2 | 6 - 7 | 2418 | 103 | ||
GenBank mRNA | BT133312.1 | - | 6 - 7 | 2410 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |