Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RluA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201853.2 | NP_001188782.2 | 3 - 4 | 578 | 89 | |
NM_001201854.2 | NP_001188783.1 | 2 - 3 | 711 | 89 | ||
NM_001273423.1 | NP_001260352.1 | 2 - 3 | 711 | 89 | ||
NM_135567.4 | NP_609411.1 | 2 - 3 | 711 | 89 | ||
GenBank mRNA | AY058686.1 | - | 2 - 3 | 663 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |