Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259171.2 | NP_001246100.1 | 5 - 6 | 1602 | 56 | |
NM_057870.4 | NP_477218.1 | 6 - 7 | 1394 | 56 | ||
NM_057871.4 | NP_477219.1 | 6 - 7 | 1520 | 56 | ||
NM_165318.3 | NP_724241.1 | 5 - 6 | 1455 | 56 | ||
GenBank mRNA | AB007821.1 | - | 6 - 7 | 1432 | 56 | |
AB007822.1 | - | 6 - 7 | 1381 | 56 | ||
AY070549.1 | - | 6 - 7 | 1349 | 56 | ||
BT120173.1 | - | 5 - 6 | 1569 | 56 | ||
BT120181.1 | - | 5 - 6 | 1569 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |