Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273746.2 | NP_001260675.1 | 3 - 4 | 800 | 61 | |
NM_136264.6 | NP_610108.1 | 3 - 4 | 660 | 61 | ||
NM_165378.1 | NP_724339.1 | 3 - 4 | 816 | 61 | ||
NM_165379.3 | NP_724340.1 | 3 - 4 | 683 | 61 | ||
GenBank mRNA | AF202633.1 | - | 3 - 4 | 580 | 61 | |
AY314744.1 | - | 3 - 4 | 587 | 61 | ||
BT044501.1 | - | 3 - 4 | 816 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |