Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273746.2 | NP_001260675.1 | 6 - 7 | 1082 | 97 | |
NM_136264.6 | NP_610108.1 | 5 - 6 | 936 | 97 | ||
NM_165378.1 | NP_724339.1 | 5 - 6 | 1092 | 97 | ||
NM_165379.3 | NP_724340.1 | 5 - 6 | 959 | 97 | ||
GenBank mRNA | AF202633.1 | - | 5 - 6 | 856 | 97 | |
AY314744.1 | - | 5 - 6 | 863 | 97 | ||
BT044501.1 | - | 5 - 6 | 1092 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |