Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Df31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032217.3 | NP_001027388.1 | 2 - 3 | 640 | 95 | |
NM_078890.4 | NP_523614.1 | 2 - 3 | 640 | 95 | ||
NM_165388.4 | NP_724350.1 | 3 - 4 | 532 | 95 | ||
GenBank mRNA | AF005846.1 | - | 2 - 3 | 765 | 95 | |
AY051664.1 | - | 2 - 3 | 605 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |