Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299285.1 | NP_001286214.1 | 3 - 4 | 1306 | 81 | |
NM_001299286.1 | NP_001286215.1 | 3 - 4 | 1306 | 81 | ||
NM_136570.4 | NP_610414.1 | 3 - 4 | 1306 | 81 | ||
NM_165630.3 | NP_724716.1 | 3 - 4 | 1306 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |