Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ork1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298167.1 | NP_001285096.1 | 6 - 7 | 1885 | 81 | |
NM_078557.5 | NP_511112.1 | 6 - 7 | 1455 | 81 | ||
NM_167259.3 | NP_727466.1 | 6 - 7 | 1576 | 81 | ||
GenBank mRNA | BT044148.1 | - | 6 - 7 | 1456 | 81 | |
U55321.1 | - | 6 - 7 | 1132 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster Inventoried |