Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103497.2 | NP_001096967.1 | 6 - 7 | 2273 | 81 | |
NM_078589.4 | NP_511144.2 | 5 - 6 | 1955 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |