Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201664.2 | NP_001188593.1 | 8 - 9 | 3459 | 75 | |
NM_001272635.2 | NP_001259564.1 | 8 - 9 | 3004 | 75 | ||
NM_001272637.2 | NP_001259566.1 | 8 - 9 | 3516 | 75 | ||
NM_078605.3 | NP_511160.2 | 8 - 9 | 3920 | 75 | ||
GenBank mRNA | BT132669.1 | - | 7 - 8 | 3585 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |