Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078768.4 | NP_523492.1 | 1 - 2 | 529 | 77 | |
GenBank mRNA | AY047539.1 | - | 1 - 2 | 493 | 77 | |
L08501.1 | - | 1 - 2 | 450 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |