Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sano | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144234.2 | NP_001137706.1 | 6 - 7 | 3703 | 89 | |
NM_001299666.1 | NP_001286595.1 | 4 - 5 | 3368 | 89 | ||
NM_079064.4 | NP_523788.2 | 4 - 5 | 3368 | 89 | ||
NM_166314.3 | NP_725838.1 | 6 - 7 | 3447 | 89 | ||
NM_166315.3 | NP_725839.1 | 7 - 8 | 3750 | 89 | ||
NM_166316.3 | NP_725840.1 | 4 - 5 | 3602 | 89 | ||
GenBank mRNA | BT012302.1 | - | 7 - 8 | 3747 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |