Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299747.1 | NP_001286676.1 | 7 - 8 | 3504 | 86 | |
NM_080094.4 | NP_524833.2 | 6 - 7 | 3399 | 86 | ||
NM_166446.3 | NP_726065.1 | 6 - 7 | 3399 | 86 | ||
NM_176244.3 | NP_788424.1 | 6 - 7 | 3399 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |